CH6371378945 Chem Impex International, Inc.

Boc-L-Phe(4-NHZ)-OH�A

  • MDL number MFCD08236868

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-155
Synonym Boc-4-(Z-amino)-L-phenylalanine dicyclohexylammonium salt
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-phe-4-nhz-oh-font-size-2-face-symbol-middot-font-dcha/3378?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-phe-4-nhz-oh-font-size-2-face-symbol-middot-font-dcha/3378?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart