CH6371378946 Chem Impex International, Inc.

Boc-3,4-difluoro-L-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD01075089
  • Cas Number : 198474-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 192-198
Synonym Boc-L-Phe(3,4-DiF)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-4-difluoro-l-phenylalanine-dicyclohexylammonium-salt/3379?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-4-difluoro-l-phenylalanine-dicyclohexylammonium-salt/3379?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart