CH6371378949 Chem Impex International, Inc.

Fmoc-pentafluoro-D-phenylalanine

  • MDL number MFCD00672558
  • Cas Number : 198545-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Phe(F)5-OH
Purity 0.98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-pentafluoro-d-phenylalanine/3382?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-pentafluoro-d-phenylalanine/3382?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart