CH6371378956 Chem Impex International, Inc.

Fmoc-Pal-Linker

  • MDL number MFCD00236731
  • Cas Number : 115109-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at = -4 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-pal-linker/3412?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-pal-linker/3412?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart