CH6371378960 Chem Impex International, Inc.

Boc-Ala-Pro-OH

  • MDL number MFCD00134853
  • Cas Number : 33300-72-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-160
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-ala-pro-oh/3417?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-ala-pro-oh/3417?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart