CH6371378961 Chem Impex International, Inc.

Boc-Leu-Leu-OMe

  • MDL number MFCD00235764
  • Cas Number : 15136-12-6
  • Molecular Weight : 358.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-141
Purity 99
Storage temp. Store at 0-8
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/boc-leu-leu-ome/3419?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-leu-leu-ome/3419?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart