CH6371378964 Chem Impex International, Inc.

Boc-Phe-Leu-OH

  • MDL number MFCD00190821
  • Cas Number : 33014-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.5
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-phe-leu-oh/3423?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-phe-leu-oh/3423?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart