CH6371378965 Chem Impex International, Inc.

Boc-Val-Pro-OH

  • MDL number MFCD00076997
  • Cas Number : 23361-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-valyl proline
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-val-pro-oh/3425?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-val-pro-oh/3425?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart