CH6371378968 Chem Impex International, Inc.

L-meta-Tyrosine

  • MDL number MFCD00063059
  • Cas Number : 587-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym m-L-Tyr-OH
Purity 100
Storage temp. Store at 0-8
Physical Form Pale brown powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-meta-tyrosine/3428?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-meta-tyrosine/3428?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart