CH6371378969 Chem Impex International, Inc.

L-2-Hydroxyphenylalanine

  • MDL number MFCD01632033
  • Cas Number : 7423-92-9
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235 - 246 °C (dec.)
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Pale yellow solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-2-hydroxyphenylalanine/3429?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-2-hydroxyphenylalanine/3429?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart