CH6371378973 Chem Impex International, Inc.

Boc-4-(aminomethyl)-L-phenylalanine

  • MDL number MFCD01318277
  • Cas Number : 137452-49-4
  • Molecular Weight : 294.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8 °C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-4-aminomethyl-l-phenylalanine/3433?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-4-aminomethyl-l-phenylalanine/3433?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart