CH6371378987 Chem Impex International, Inc.

Boc-S-benzyl-D-cysteine

  • MDL number MFCD00038253
  • Cas Number : 102830-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-87
Synonym Boc-D-Cys(Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-d-cysteine/3448?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-benzyl-d-cysteine/3448?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart