CH6371378996 Chem Impex International, Inc.

Na-Fmoc-Nd-2-chloro-Z-L-ornithine

  • MDL number MFCD00235825
  • Cas Number : 198561-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Orn(2-Cl-Z)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-delta-sup-2-chloro-z-l-ornithine/3460?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-delta-sup-2-chloro-z-l-ornithine/3460?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart