CH6371379002 Chem Impex International, Inc.

Z-N-methylglycine N-hydroxysuccinimide ester

  • MDL number MFCD00153340
  • Cas Number : 53733-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-sarcosine N-hydroxysuccinimide ester
Purity 97
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-n-i-methylglycine-i-n-i-hydroxysuccinimide-ester/3466?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-n-i-methylglycine-i-n-i-hydroxysuccinimide-ester/3466?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart