CH6371379007 Chem Impex International, Inc.

Fmoc-O-benzyl-D-tyrosine

  • MDL number MFCD00235837
  • Cas Number : 138775-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-142
Synonym Fmoc-D-Tyr(Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-benzyl-d-tyrosine/3471?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-benzyl-d-tyrosine/3471?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart