CH6371379008 Chem Impex International, Inc.

Boc-L-valine 4-nitrophenyl ester

  • MDL number MFCD00076995
  • Cas Number : 16948-40-6
  • Molecular Weight : 338.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-69 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-valine-4-nitrophenyl-ester/3472?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-valine-4-nitrophenyl-ester/3472?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart