CH6371379011 Chem Impex International, Inc.

3-(4-Hydroxymethylphenoxy)propionic acid

  • MDL number MFCD00137851
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[4-(hydroxymethyl)phenoxy]propanoic acid
Inchi Code 1S/C10H12O4/c11-7-8-1-3-9(4-2-8)14-6-5-10(12)13/h1-4,11H,5-7H2,(H,12,13)
Inchi Key WPWKJUVXEMKOHS-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/3-4-hydroxymethylphenoxy-propionic-acid/3485?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-4-hydroxymethylphenoxy-propionic-acid/3485?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart