CH6371379013 Chem Impex International, Inc.

Boc-S-4-methylbenzyl-L-penicillamine dicyclohexylammonium salt

  • MDL number MFCD00235907
  • Cas Number : 198474-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-l-penicillamine-dicyclohexylammonium-salt/3487?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methylbenzyl-l-penicillamine-dicyclohexylammonium-salt/3487?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart