CH6371379014 Chem Impex International, Inc.

D-Thiaproline

  • MDL number MFCD00137425
  • Cas Number : 45521-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-193 �dec.)
Synonym D-Thz-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-thiaproline/3488?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-thiaproline/3488?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart