CH6371379015 Chem Impex International, Inc.

Fmoc-3-iodo-L-tyrosine

  • MDL number MFCD00235906
  • Cas Number : 134486-00-3
  • Molecular Weight : 529.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.5
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-3-iodo-l-tyrosine/3489?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-3-iodo-l-tyrosine/3489?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart