CH6371379016 Chem Impex International, Inc.

Na-Fmoc-Nim-4-toluenesylfonyl-L-histidine

  • MDL number MFCD00144353
  • Cas Number : 112380-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-His(Tos)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-im-sup-4-toluenesylfonyl-l-histidine/3490?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-im-sup-4-toluenesylfonyl-l-histidine/3490?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart