CH6371379017 Chem Impex International, Inc.

Fmoc-D-threonine

  • MDL number MFCD00153365
  • Cas Number : 118609-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 83-90
Synonym Fmoc-D-Thr-OH
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-threonine/3491?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-threonine/3491?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart