CH6371379019 Chem Impex International, Inc.

Boc-S-acetamidomethyl-D-cysteine

  • MDL number MFCD00076917
  • Cas Number : 138775-00-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-116� C
Synonym Boc-D-Cys(Acm)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-d-cysteine/3493?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-acetamidomethyl-d-cysteine/3493?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart