CH6371379023 Chem Impex International, Inc.

Z-D-norleucine

  • MDL number MFCD00063169
  • Cas Number : 15027-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-65
Synonym Z-D-Nle-OH
Purity 0.99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-d-norleucine/3497?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-d-norleucine/3497?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart