CH6371379027 Chem Impex International, Inc.

Ne-Fmoc-L-lysine methyl ester hydrochloride

  • MDL number MFCD00153447
  • Cas Number : 201009-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163-167
Synonym L-Lys(Fmoc)-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-e-sup-fmoc-l-lysine-methyl-ester-hydrochloride/3501?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-e-sup-fmoc-l-lysine-methyl-ester-hydrochloride/3501?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart