CH6371379036 Chem Impex International, Inc.

Na-[2,4-Dinitro-5-fluorophenyl]-D-alanine amide

  • MDL number MFCD00155642
  • Cas Number : 132055-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym FDNP-D-Ala-NH2
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-2-4-dinitro-5-fluorophenyl-d-alanine-amide/3510?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-2-4-dinitro-5-fluorophenyl-d-alanine-amide/3510?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart