CH6371379039 Chem Impex International, Inc.

Na-Boc-Ne-formyl-L-lysine

  • MDL number MFCD00037361
  • Cas Number : 2483-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-128
Synonym Boc-L-Lys(For)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-formyl-l-lysine/3514?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-formyl-l-lysine/3514?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart