CH6371379040 Chem Impex International, Inc.

Boc-S-methyl-L-penicillamine dicyclohexylammonium salt

  • MDL number MFCD00070261
  • Cas Number : 112898-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Pen(Me)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-methyl-l-penicillamine-dicyclohexylammonium-salt/3515?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-methyl-l-penicillamine-dicyclohexylammonium-salt/3515?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart