CH6371379046 Chem Impex International, Inc.

Glycine benzyloxycarbonylhydrazide trifluoroacetate

  • MDL number MFCD00058603
  • Cas Number : 19704-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-181� C
Synonym Gly-NHNHZ TFA
Purity 98
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/glycine-benzyloxycarbonylhydrazide-trifluoroacetate/3522?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/glycine-benzyloxycarbonylhydrazide-trifluoroacetate/3522?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart