CH6371379047 Chem Impex International, Inc.

Leucine benzyloxycarbonylhydrazide trifluoroacetate

  • MDL number MFCD00058483
  • Cas Number : 19635-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-186
Synonym L-Leu-NHNHZ TFA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/leucine-benzyloxycarbonylhydrazide-trifluoroacetate/3523?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/leucine-benzyloxycarbonylhydrazide-trifluoroacetate/3523?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart