CH6371379056 Chem Impex International, Inc.

Boc-O-2',6'-dichlorobenzyl-3,5-diiodo-L-tyrosine

  • MDL number MFCD00076994
  • Cas Number : 201416-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186 - 191 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-3-5-diiodo-l-tyrosine/3540?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-2-6-dichlorobenzyl-3-5-diiodo-l-tyrosine/3540?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart