CH6371379057 Chem Impex International, Inc.

Boc-3,5-diiodo-L-tyrosine methyl ester

  • MDL number MFCD00237523
  • Cas Number : 128781-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-123
Synonym Boc-3,5-diiodo-L-Tyr-OMe
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-3-5-diiodo-l-tyrosine-methyl-ester/3541?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-3-5-diiodo-l-tyrosine-methyl-ester/3541?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart