CH6371379061 Chem Impex International, Inc.

Nalpha-Boc-Nomega-(methoxybenzenesulfonyl)-L-arginine

  • MDL number MFCD00134857
  • Cas Number : 58810-09-6
  • Molecular Weight : 444.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-149 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-omega-sup-methoxybenzenesulfonyl-l-arginine/3546?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-omega-sup-methoxybenzenesulfonyl-l-arginine/3546?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart