CH6371379062 Chem Impex International, Inc.

Na-Boc-Nw-(mesitylene-2-sulfonyl)-L-arginine cyclohexylammonium salt

  • MDL number MFCD00065555
  • Cas Number : 68262-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Arg(Mts)-OH CHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-sup-mesitylene-2-sulfonyl-l-arginine-cyclohexylammonium-salt/3547?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-sup-mesitylene-2-sulfonyl-l-arginine-cyclohexylammonium-salt/3547?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart