CH6371379066 Chem Impex International, Inc.

Nalpha-Boc-L-asparagine N-hydroxysuccinimide ester

  • MDL number MFCD00190785
  • Cas Number : 42002-18-6
  • Molecular Weight : 329.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-asparagine-i-n-i-hydroxysuccinimide-ester/3552?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-asparagine-i-n-i-hydroxysuccinimide-ester/3552?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart