CH6371379071 Chem Impex International, Inc.

Boc-S-ethyl-L-cysteine

  • MDL number MFCD00076919
  • Cas Number : 16947-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-60� C
Synonym Boc-L-Cys(Et)-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-ethyl-l-cysteine/3558?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-ethyl-l-cysteine/3558?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart