CH6371379074 Chem Impex International, Inc.

Boc-S-Z-aminoethyl-L-cysteine

  • MDL number MFCD00236840
  • Cas Number : 85003-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Cys(Z-NHEt)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Clear, colorless oil
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-z-aminoethyl-l-cysteine/3561?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-z-aminoethyl-l-cysteine/3561?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart