CH6371379076 Chem Impex International, Inc.

Na-Boc-Nw-(4-methoxy-2,3,6-trimethylbenzenesulfonyl)-D-arginine

  • MDL number MFCD00151863
  • Cas Number : 200122-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92-97
Synonym Boc-D-Arg(Mtr)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-omega-sup-4-methoxy-2-3-6-trimethylbenzenesulfonyl-d-arginine/3563?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-omega-sup-4-methoxy-2-3-6-trimethylbenzenesulfonyl-d-arginine/3563?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart