CH6371379077 Chem Impex International, Inc.

Na-Boc-Nw,w'-bis-Z-D-arginine

  • MDL number MFCD00236825
  • Cas Number : 145881-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137-141 �C
Synonym Boc-D-Arg(Z)2-OH
Purity 99
Storage temp. Store at = -4
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-omega-omega-sup-bis-z-d-arginine/3565?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-omega-omega-sup-bis-z-d-arginine/3565?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart