CH6371379078 Chem Impex International, Inc.

Nalpha-Boc-Ngamma-xanthyl-D-asparagine

  • MDL number MFCD00076910
  • Cas Number : 200192-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-184 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-gamma-sup-xanthyl-d-asparagine/3566?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-gamma-sup-xanthyl-d-asparagine/3566?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart