CH6371379082 Chem Impex International, Inc.

Boc-D-aspartic acid a-tert-butyl ester

  • MDL number MFCD00236827
  • Cas Number : 77004-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Asp-OtBu
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-aspartic-acid-a-i-tert-i-butyl-ester/3571?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-aspartic-acid-a-i-tert-i-butyl-ester/3571?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart