CH6371379084 Chem Impex International, Inc.

Boc-S-4-methoxybenzyl-D-cysteine

  • MDL number MFCD00076922
  • Cas Number : 58290-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-74
Synonym Boc-D-Cys(pMeOBzl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methoxybenzyl-d-cysteine/3574?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-4-methoxybenzyl-d-cysteine/3574?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart