CH6371379086 Chem Impex International, Inc.

Nalpha-Boc-Ndelta-xanthyl-D-glutamine

  • MDL number MFCD00151873
  • Cas Number : 99092-88-3
  • Molecular Weight : 426.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-150 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-delta-sup-xanthyl-d-glutamine/3576?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-delta-sup-xanthyl-d-glutamine/3576?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart