CH6371379087 Chem Impex International, Inc.

Boc-D-glutamic acid g-cyclohexyl ester

  • MDL number MFCD00076929
  • Cas Number : 133464-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Glu(OcHex)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-cyclohexyl-ester/3577?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-cyclohexyl-ester/3577?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart