CH6371379088 Chem Impex International, Inc.

Boc-D-glutamic acid gamma-9-fluorenylmethyl ester

  • MDL number MFCD00151871
  • Cas Number : 123417-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115 - 121 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-9-fluorenylmethyl-ester/3578?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-9-fluorenylmethyl-ester/3578?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart