CH6371379091 Chem Impex International, Inc.

Boc-D-glutamic acid alpha-9-fluorenylmethyl ester

  • MDL number MFCD00798626
  • Cas Number : 214630-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-102° C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-a-9-fluorenylmethyl-ester/3581?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-a-9-fluorenylmethyl-ester/3581?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart