CH6371379094 Chem Impex International, Inc.

Boc-D-glutamic acid a-tert-butyl ester

  • MDL number MFCD00076926
  • Cas Number : 73872-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-Glu-OtBu
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-alpha-i-tert-i-butyl-ester/3584?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-glutamic-acid-alpha-i-tert-i-butyl-ester/3584?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart