CH6371379096 Chem Impex International, Inc.

Na-Boc-Nim-benzyl-D-histidine

  • MDL number MFCD00038628
  • Cas Number : 65717-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-184�
Synonym Boc-D-His(Bzl)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-im-sup-benzyl-d-histidine/3586?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-im-sup-benzyl-d-histidine/3586?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart