CH6371379097 Chem Impex International, Inc.

Na-Boc-Nim-2,4-dinitrophenyl-D-histidine�propanol

  • MDL number MFCD00672320

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-D-His(Dnp)-OH IPA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-im-sup-2-4-dinitrophenyl-d-histidine·isopropanol/3587?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-i-n-i-sup-im-sup-2-4-dinitrophenyl-d-histidine·isopropanol/3587?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart