CH6371379101 Chem Impex International, Inc.

Boc-D-aspartic acid alpha-amide

  • MDL number MFCD00151866
  • Cas Number : 200282-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103 - 109 °C
Purity 98
Storage temp. Store at 0 - 8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-aspartic-acid-a-amide/3593?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-aspartic-acid-a-amide/3593?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart